missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Guanidine Hydrochloride, ∽99%, (Crystalline/Optical Grade), MP Biomedicals

Produces clear, colorless solutions

$74.39 - $666.65

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15121025
View Documents
MP Biomedicals Inc
0215121025
25 g
Each for $74.39
 
ICN15121090
View Documents
MP Biomedicals Inc
0215121090
500 g
Each for $372.26
 
ICN15121091
View Documents
MP Biomedicals Inc
0215121091
1 kg
Each for $666.65
 
Description

Description

Specifications

Chemical Identifiers

50-01-1
95.53
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
CHEBI:32735
C(=N)(N)N.Cl
CH6ClN3
PJJJBBJSCAKJQF-UHFFFAOYSA-N
5742
guanidine;hydrochloride

Specifications

50-01-1
CH6ClN3
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
C(=N)(N)N.Cl
95.53
CHEBI:32735
Optical
Guanidine Hydrochloride
∽99%
25 g
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine;hydrochloride
5742
95.5
Crystalline
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'