Learn More
Guanine, 98%, Thermo Scientific Chemicals
$52.85 - $494.10
Chemical Identifiers
CAS | 73-40-5 |
---|---|
Molecular Formula | C5H5N5O |
Molecular Weight (g/mol) | 151.13 |
MDL Number | MFCD00071533 |
InChI Key | UYTPUPDQBNUYGX-UHFFFAOYSA-N |
Synonym | guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence |
PubChem CID | 764 |
ChEBI | CHEBI:16235 |
SMILES | NC1=NC(=O)C2=C(N1)N=CN2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1202414
|
Thermo Scientific Chemicals
A1202414 |
25 g |
Each for $52.85
|
|
|||||
AAA1202422
|
Thermo Scientific Chemicals
A1202422 |
100 g |
Each for $143.16
|
|
|||||
AAA1202436
|
Thermo Scientific Chemicals
A1202436 |
500 g |
Each for $494.10
|
|
|||||
Description
Employed as a nucleobase incorporated into biological nucleic acids. Guanine is suitable reagent used to investigate the mechanism of electrochemical oxidation of guanine and adenine using a glassy carbon microelectrode and cyclic and differential pulse voltammetry. It may be used in the preparation of mixed-sequence peptide nucleic acid (PNA) monomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as a nucleobase incorporated into biological nucleic acids. Guanine is suitable reagent used to investigate the mechanism of electrochemical oxidation of guanine and adenine using a glassy carbon microelectrode and cyclic and differential pulse voltammetry. It may be used in the preparation of mixed-sequence peptide nucleic acid (PNA) monomers.
Solubility
Insoluble in water. Soluble in ammonia-water, KOH solutions, dilute acids, and 1 N NaOH (0.1M).
Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
Chemical Identifiers
73-40-5 | |
151.13 | |
UYTPUPDQBNUYGX-UHFFFAOYSA-N | |
764 | |
NC1=NC(=O)C2=C(N1)N=CN2 |
C5H5N5O | |
MFCD00071533 | |
guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence | |
CHEBI:16235 |
Specifications
73-40-5 | |
C5H5N5O | |
25 g | |
14,4564 | |
Insoluble in water. Soluble in ammonia-water,KOH solutions,dilute acids,and 1 N NaOH (0.1M). | |
NC1=NC(=O)C2=C(N1)N=CN2 | |
151.13 | |
CHEBI:16235 | |
98% |
>300°C | |
MFCD00071533 | |
9680 | |
guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence | |
UYTPUPDQBNUYGX-UHFFFAOYSA-N | |
2-amino-6,7-dihydro-3H-purin-6-one | |
764 | |
151.13 | |
Guanine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-799-8
RTECSNumber : MF8260000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only