missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine 5'-monophosphate, disodium salt hydrate, 97%
CAS: 5550-12-9 | C10H12N5Na2O8P | 407.19 g/mol
$154.38 - $606.60
Chemical Identifiers
| CAS | 5550-12-9 |
|---|---|
| Molecular Formula | C10H12N5Na2O8P |
| Molecular Weight (g/mol) | 407.19 |
| MDL Number | MFCD00151238 |
| InChI Key | PVBRXXAAPNGWGE-KHRSEZDTNA-L |
| PubChem CID | 21712 |
| IUPAC Name | disodium;[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate |
| SMILES | [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC226600050
|
Thermo Scientific Chemicals
226600050 |
5 g | Glass bottle |
Each for $154.38
|
|
||||
|
AC226600250
|
Thermo Scientific Chemicals
226600250 |
25 g | Glass bottle |
Each for $606.60
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5550-12-9 | |
| 407.19 | |
| PVBRXXAAPNGWGE-KHRSEZDTNA-L | |
| disodium;[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate |
| C10H12N5Na2O8P | |
| MFCD00151238 | |
| 21712 | |
| [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1 |
Specifications
| 5550-12-9 | |
| White | |
| Authentic | |
| Glass bottle | |
| MFCD00151238 | |
| 10, 279 | |
| guanosine 5'-monophosphate disodium salt, 5'-guanylic acid disodium salt, disodium guanylate, disodium 5'-guanylate, sodium guanylate, disodium gmp, disodium 5'-gmp, gmp disodium salt, 5'-guanylic acid, disodium salt, 5'-gmp disodium salt | |
| 97% min. (lambda max. 252nm,pH 7) molar absorptivity 13700+/-4%,pH 7 | |
| [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1 | |
| 407.19 | |
| 407.19 | |
| Crystalline Powder |
| >300°C | |
| 25% max. (120°C, 4 hrs) | |
| 97% | |
| C10H12N5Na2O8P | |
| 5 g | |
| 14, 4570 | |
| Solubility in water: >50g/l water | |
| PVBRXXAAPNGWGE-KHRSEZDTNA-L | |
| disodium;[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate | |
| 21712 | |
| 97% | |
| Guanosine 5′-monophosphate,disodium salt hydrate |
Safety and Handling
EINECSNumber : 226-914-1
RUO – Research Use Only