missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Guanosine 5'-triphosphate, disodium salt hydrate, 90%, for biochemistry
Guanosine 5'-triphosphate, disodium salt hydrate, 90%, C10H14N5Na2O14P3.xH2O, CAS Number-56001-37-7
Supplier: Thermo Scientific Chemicals 226250010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistrySpecifications
Guanosine 5'-triphosphate, disodium salt hydrate | |
1.12 to 1.18 (A250/A260), 0.63 to 0.69 (A280/A260) | |
White | |
88% min. (HPLC) | |
C10H14N5Na2O14P3 | |
1 g | |
Solubility in water: soluble | |
FIZIYLKEXVIRHJ-KHRSEZDTNA-L | |
disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate | |
131676145 | |
90% | |
Biochemistry |
56001-37-7 | |
180°C | |
Authentic | |
Glass bottle | |
MFCD03410297 | |
guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate | |
88% min. (lambda max. 252nm,pH 7) | |
[Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1 | |
567.14 | |
567.14 | |
4% max. (typically) Guanosine diphosphate (HPLC) | |
Crystalline Powder |
Chemical Identifiers
56001-37-7 | |
567.14 | |
FIZIYLKEXVIRHJ-KHRSEZDTNA-L | |
131676145 | |
[Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1 |
C10H14N5Na2O14P3 | |
MFCD03410297 | |
guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate | |
disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate |
Safety and Handling
EINECSNumber : 259-940-7
RUO – Research Use Only