missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Guardian™ 2000 Stirrer

Guardian™ 2000 Stirrer is ideal for education and other lab environments that require affordable, everyday reliable stirring up to 15 L. All three sizes offer simple knob control to easily adjust the stirring functions.

Supplier:  OHAUS™ 30680265

 View more versions of this product

Catalog No. 01918391


Explore available promotions

Description

Description

  • Applications: Boiling Liquids, Basic Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
  • Display: Prominent hot top indicator light, green indicator lights when unit is heating and stirring
  • Operation: Simple knob control, stirring up to 1600 rpm, temperature up to 500°C (ceramic top) or 380°C (ceramic coated stainless steel top), power cord (included), stirring models include PTFE coated stir bar
  • Communication: N/A
  • Construction: Durable metal housing, PET front panel, control knobs, ON/OFF switch for heater disconnect, built in support rod holder, easy to clean resin, ceramic or ceramic coated stainless steel tops
  • Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring
Specifications

Specifications

200 to 1600 rpm
Analog
4 x 4 in.
35 W
2.1 kg
15 L (4 gal.)
115 V
e-G21ST04R
60 Hz
4.7 lb.
Square
264 x 163 x 110 mm (10.4 x 6.4 x 4.3 in.)
Videos
SDS
Documents

Documents

Promotions

Promotions