3M™ GVP-Series Belt-Mounted Powered Air-Purifying Respirator (PAPR) Assemblies
GVP-1 PAPR Assembly offers respiratory protection and allows freedom of movement while being used in applications. Includes all the components needed to make a complete 3M PAPR System.
Supplier:  3M™ GVP1
 View more versions of this product
Catalog No. 19-087398
Explore available promotions
Alert:
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Includes:
  • Web belt or comfort belt, motor blower, battery, charger, flow meter, power cord, blower plugs
  • Comfort belt-mounted assembly includes above items but features padded CB-1000 comfort belt that adjusts from 26 to 54 in. (66 to 134cm)
Description
Description
  • For use with certain 3M respiratory headgear, filter/cartridges, batteries and breathing tubes
  • NIOSH approved respiratory system
  • Protects against certain gases, organic vapors, and particulates
  • Includes the components needed to make a complete 3M PAPR System when combined with 3M headgear and breathing tube:
    • Motor Blower
    • Flow Indicator and Adaptor
    • Blower Plugs
    • Web Belt, Urethane Coated Belt OR Comfort Belt
    • Power Coard
    • Battery Pack
    • Battery Charger
  • Single replaceable cartridge system
  • Contours to the hip and body for comfort
Specifications
Specifications
8 hours
OSHA, NIOSH
PAPR Assembly, No Headtop
PAPR respirator
GVP1
Web Belt
High Efficiency (HE)
Assembly, facility maintenance, machine operation, painting, sanding
Motor blower, battery, charger, flow meter, power cord, blower plugs
Belt-mounted
SDS
Documents
Documents
Promotions
Promotions
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product.