Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
TECHREF™ H-2358 Sample Cups/Cells, 48mm, single open ended DFS Item
Designed specifically for use with X-ray fluorescence analyzers. TECHREF™ Sample Cells are available for XRF analyzers for a variety of styles and manufacturers and are designed for ease of use.
Supplier:  TECHREF™ H2358
Numéro de catalogue. 11022101
Découvrez plus d'offres spéciales
Description
Description
  • Composed of two parts: closed-bottom cup, outer open ring to hold and seal the desired film
  • Two fill lines in the interior of cup for consistent sample size
  • Mated locking line to prevent leakage
  • Dimple on exterior floor for venting and reservoir for overflow
  • Fully disposable, does not require adapter rings and does not require disassembly to recover reusable parts
Spécifications
Spécifications
Sample Cell
Polyethylene
18.5 mm
48 mm
Single-use
ISO 9001
Carousel and single-load model Horiba (newer than SLFA-920) and Sindi XRF analyzers
Videos
FDS
Documents
Documents
Offres spéciales
Offres spéciales