Thermo Scientific™ H-6000A 6 x 1000mL Swinging-Bucket Rotor
Carry out multiple protocols in blood bank, research, biotech and pharmaceutical environments with this swinging-bucket rotor.
Supplier:  Thermo Scientific™ 11250
Catalog No. 36-101-0601
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Carry out multiple protocols in blood bank, research, biotech and pharmaceutical environments with the Thermo Scientific™ H-6000A 6 x 1000mL Swinging-Bucket Rotor.
Includes:
Rotor cover assembly (11237), rotor buckets, anodized aluminum (3, 2/pkg., 44040), rotor cover (11232), rotor locking stud (11238), balancing discs, rubber (1 pkg., 6/pkg., 00335), speed decal (11009), loctite sealing adhesive (65811), instruction manual (11243)
Warranty:
Seven years
Specifications
Specifications
H-6000A Swinging Bucket Rotor
Swinging Bucket Rotor
7,277 x g
260.6 mm
7823
90°
3,359 x g
Stainless Steel Rotor Body, Aluminum Bucket
6 x 1000 mL
5,000 rpm
120.3 mm
Thermo Scientific Sorvall RC 3 Series Large Capacity Centrifuges
Sealing lid is biocontainment certified by the Public Health Laboratory Service, Microbiology Services, Porton Down, U.K.
Yes
Videos
SDS
Documents
Documents
Accessories