SP Bel-Art™ H-B DURAC Calibrated Thermocouple Thermometers
H-B DURAC Calibrated Thermocouple Thermometers; -200/1370°C (-328/2498°F)
$904.04 - $1760.11
Specifications
Calibration Points 0°C, 100°C
Diameter (English) Probe 1.25 in.
Height (English) 7.25 in.
Height (Metric) 18.5 cm
Temperature Range (English) -328°F to +2498°F
View More Specs
Products 4
Catalog Number Mfr. No. Includes Resolution Price Quantity  
Catalog Number Mfr. No. Includes Resolution Price Quantity  
01-257-124
View Documents
N/A 2 Probe Inputs/ Displays, RS232 Connection K-type: 0.1°C between -200/800°C, otherwise 1°CJ-type: 0.1°C between -200/600°C, otherwise 1°C
Each for $904.04
Case of 3 Each for $2,712.12
01-257-125
View Documents
N/A 4 Probe Inputs/ Displays, RS232 Connection 0.1°C between -200/200°C, otherwise 1°C
Each for $1,100.04
Case of 3 Each for $3,300.12
01-257-126
View Documents
N/A 4 Probe Inputs/ Displays, Data Logger 16,000 Points / Channel, USB Connection for Data Export, Foldaway Stand, Auto-Off, Instant Recall, Software CD 0.1°C between -200/600°C, otherwise 1°C
Each for $1,466.79
Case of 3 Each for $4,400.37
01-257-127
View Documents
N/A 4 Probe Inputs/ Displays, Data Logger 32,000 Points / Channel, Bluetooth or USB Connection for Data Export, Foldaway Stand, Auto-Off, Instant Recall, Software CD J-,K-, E-, T-, N-type: 0.1°C between -200/600°C, otherwise 1°CR-, S-type: 0.2°C between -200/600°C, otherwise 1°C
Each for $1,760.11
Case of 3 Each for $5,280.33
Description
Description
  • Used for temperature measurement in life science research, kilns, gas turbine exhaust, diesel engines, and other industrial processes, H-B Calibrated Thermocouple Thermometers provide measurement throughout a broad temperature range and the ability to utilize multiple probes at one time.
  • Wide temperature range allows accurate monitoring at extreme temperatures
  • Max/min/average temperature recording capability
  • Data hold
  • Includes two K-type beaded wire probes (-20/250°C capability); one is calibrated and serialized
  • Software CD (where included) for real time data collection and logging via USB
  • Bluetooth™ app available for use with B60911-0200
  • Probes for wider temperature ranges available (B60901-1200 & B60901-1300) or use alternatively purchased probes; see chart for probe types accepted by each unit 
  • Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
  • Individually serialized, CE-marked Note: calibration applies to included probe and tested port only
Specifications
Specifications
0°C, 100°C
7.25 in.
-328°F to +2498°F
1.25 in.
18.5 cm
-200°C to +1370°C
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'