missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SP Bel-Art™ H-B DURAC Thermocouple Thermometer Probes

H-B DURAC Thermocouple Thermometer Probe

$507.64 - $528.96

Specifications

Length (English) Cable 40 in.
Length (Metric) Cable 101.5 cm
View More Specs

Products 2
Catalog Number Mfr. No. Probe Type Temperature Range (Metric) Price Quantity  
Catalog Number Mfr. No. Probe Type Temperature Range (Metric) Price Quantity  
01-257-128
View Documents
N/A J, lead wire, stainless steel overbraid -40°C to +750°C
Each for $528.96
 
01-257-129
View Documents
N/A K, lead wire, FEP-insulated -200°C to +920°C
Each for $507.64
 
Description

Description

  • For use with H-B DURAC Calibrated Thermocouple Thermometers or others that accept K or J type probes.
  • For temperature measurement of liquids, semi-solids, or air/gasses
  • Measure wide temperature ranges
  • 15.24 cm (6”) stainless steel probe
  • B60901-1300 perfect for use in dewars
Specifications

Specifications

40 in.
101.5 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'