missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm3) Plain Form Hydrometers: for Liquids Heavier Than Water

Available in a variety of models measuring specific gravity/relative density (g/cm3) of liquids heavier than water in various ranges.

$99.75 - $139.50

Specifications

Certifications/Compliance Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory (Cert 2448.01); internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.
Content And Storage Plain form hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
For Use With (Application) For Liquids Heavier Than Water
View More Specs

Products 6
Catalog Number Mfr. No. Specific Gravity Range Accuracy Increments Length (Metric) Price Quantity  
Catalog Number Mfr. No. Specific Gravity Range Accuracy Increments Length (Metric) Price Quantity  
13-202-421
View Documents
H-B Instrument™
B618017200RMD
1.000 to 1.220 ±0.002 0.002 300 mm
Each for $99.75
 
13-202-098
View Documents
H-B Instrument™
B618014800
1.000 to 1.250 ±0.005 0.005 165 mm
Each for $101.15
 
13-202-422
View Documents
H-B Instrument™
B618017300RMD
1.200 to 1.420 ±0.002 0.002 300 mm
Each for $111.35
 
13-202-118
View Documents
N/A 1.000 to 1.600 ±0.005 0.005 300 mm
Each for $117.57
 
13-202-099
View Documents
H-B Instrument™
B618014900
1.200 to 1.450 ±0.005 0.005 165 mm
Each for $103.12
 
13-202-423
View Documents
N/A 1.400 to 1.620 ±0.002 0.002 300 mm
Each for $139.50
 
Description

Description

Available in a variety of models measuring specific gravity/relative density (g/cm3) of liquids heavier than water in various ranges.

  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60°F (15°C)
Applications:
Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.

Certifications:
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory (Cert 2448.01); internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.

Storage:

Plain form hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.

Order Info

Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Specifications

Specifications

Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory (Cert 2448.01); internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.
Plain form hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
For Liquids Heavier Than Water
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'