Learn More
Thermo Scientific™ H-Flex 1  Blood Tube Package
H-Flex 1 rotor blood tube package offers the flexibility to tun microplates, 5/7 mL and 10/12 mL blood tubes in the same rotor.
Supplier:  Thermo Scientific™ 75000156
Catalog No. 75000156
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
H-Flex 1 rotor blood processing package containing adapters for microplates, 5/7 ml and 10/12 ml blood tubes
  • H-Flex 1 rotor blood processing package containing adapters for microplates, 5/7 mL and 10/12 mL blood tubes
Specifications
Specifications
Buckets are available with biocontainment certified lids
2x10 standard microplates (2x1 deepwell), 66 x 10/12 mL 84 x 5/7 mL blood tubes
4297
  • 75003300 H-Flex 1 Rotor
  • 75003301 H-Flex 1 Buckets
  • 75003302 H-Flex 1 Lids
  • 75003307 Set of 2 Microplate Carriers, 5 standard, 1 deepwell
  • 75003305 Set of 2 adapters, 33 x 10/12 ml blood tubes
  • 75003306 Set of 2 adapters 42 x 5/7 ml blood tubes
Buckets are available with biocontainment certified lids
4,700 x g
Sorvall
Can be used with Thermo Scientific Sorvall Legend X1, X1 Pro and ST16, ST1 Plus Series, Thermo Scientific Multifuge X1, X1 Pro and Megafuge 16, ST1 Plus Series, Thermo Scientific SL 16, ST1 Plus Series
Product Suggestions
Product Suggestions