Honeywell Miller™ H500 Series Safety Harness, Model IC5
The modern ergonomic design provides high visibility and smart features, in addition to safe and comfortable functionality. Features highly breathable,lightweight, ergonomic shoulder/back padding.
Supplier: Honeywell Miller™ H5IC311123
Description
- Lightweight, ergonomic, breathable harness
- Comfortable around the legs, waist, back and shoulders
- Smart rebar hook/lanyard keepers and SRL web link
- Increased freedom of movement
- Quick and easy-to-fit and adjust
- Improved visible stitch pattern
- Clearly identifiable fall-impact indicators
Specifications
Tongue/Mating | |
2X-Large | |
Steel |
Fall Protection Harness | |
H500 IC5 XXL STL TB/MB FD/SD | |
Front and Side |