missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hafnium arc-cast pellet, 12mm (0.47 in.) dia., 99.8% (metals basis excluding Zr), Zr nominal 4.5%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 45104KF
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Hafnium arc-cast pellet | |
2227°C | |
4603°C | |
(metals basis excluding Zr), Zr nominal 4.5% | |
MFCD00011032 | |
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire | |
[Hf] | |
178.49 | |
CHEBI:33343 | |
99.8% | |
≈10g/pellet |
12mm (0.47 in.) dia. | |
7440-58-6 | |
13.310 g/mL | |
Hf | |
1 Pc. | |
VBJZVLUMGGDVMO-UHFFFAOYSA-N | |
hafnium | |
23986 | |
178.49 | |
Odorless | |
Pellet |
Chemical Identifiers
7440-58-6 | |
178.49 | |
VBJZVLUMGGDVMO-UHFFFAOYSA-N | |
23986 | |
hafnium |
Hf | |
MFCD00011032 | |
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire | |
CHEBI:33343 | |
[Hf] |
Safety and Handling
EINECSNumber : 231-166-4
TSCA : No
Recommended Storage : Ambient temperatures