Learn More
Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals
$35.55 - $411.27
Chemical Identifiers
CAS | 13499-05-3 |
---|---|
Molecular Formula | Cl4Hf |
Molecular Weight (g/mol) | 320.29 |
MDL Number | MFCD00011034 |
InChI Key | PDPJQWYGJJBYLF-UHFFFAOYSA-J |
Synonym | hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis |
PubChem CID | 37715 |
IUPAC Name | tetrachlorohafnium |
SMILES | Cl[Hf](Cl)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1183488
|
Thermo Scientific Chemicals
01183488 |
2.5 g |
Each for $35.55
|
|
|||||
AA1183414
|
Thermo Scientific Chemicals
01183414 |
25 g |
Each for $119.99
|
|
|||||
AA1183422
|
Thermo Scientific Chemicals
01183422 |
100 g |
Each for $411.27
|
|
|||||
Description
Hafnium chloride tetrahydrofuran complex is an efficient catalyst for the Diels-Alder cycloadditions, Ziegler-Natta polymerization of alkenes. It is used as control rods in nuclear reactors and in manufacture of light bulb filaments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13499-05-3 | |
320.29 | |
PDPJQWYGJJBYLF-UHFFFAOYSA-J | |
37715 | |
Cl[Hf](Cl)(Cl)Cl |
Cl4Hf | |
MFCD00011034 | |
hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis | |
tetrachlorohafnium |
Specifications
320° (sublimation) | |
White | |
2.5 g | |
Cl4Hf | |
hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis | |
PDPJQWYGJJBYLF-UHFFFAOYSA-J | |
tetrachlorohafnium | |
37715 | |
Irritating | |
Hafnium(IV) chloride |
13499-05-3 | |
Powder | |
>98% (Metals basis excluding Zr) | |
MFCD00011034 | |
Soluble in water,methanol and acetone. | |
Cl[Hf](Cl)(Cl)Cl | |
320.29 | |
320.3 | |
Moisture sensitive |
Safety and Handling
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : DOT Class: 8; Packaging Group: III
EINECSNumber : 236-826-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only