missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hafnium powder, -60+325 mesh, 99.6% (metals basis excluding Zr), Zr nominal 2-3.5%, Thermo Scientific Chemicals

$190.23 - $190.23

Chemical Identifiers

CAS 7440-58-6
Molecular Formula Hf
Molecular Weight (g/mol) 178.49
MDL Number MFCD00011032
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
Synonym hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
PubChem CID 23986
ChEBI CHEBI:33343
IUPAC Name hafnium
SMILES [Hf]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1020209
View Documents
Thermo Scientific Chemicals
01020209
10 g
Each for $190.23
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-58-6
178.49
VBJZVLUMGGDVMO-UHFFFAOYSA-N
23986
hafnium
Hf
MFCD00011032
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
CHEBI:33343
[Hf]

Specifications

Gray
Hf
UN2545
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
[Hf]
178.49
CHEBI:33343
−60+325 Mesh
Powder
(metals basis excluding Zr), Zr nominal 2 to 3.5%
7440-58-6
MFCD00011032
10 g
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium
23986
99.6%
Odorless
Hafnium powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H251
Self-heating: may catch fire.

P210-P222-P280-P335+P334-P370+P378q

H250

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: HAFNIUM POWDER, DRY

EINECSNumber : 231-166-4

RTECSNumber : MG4600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only