missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hafnium trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

$193.84 - $604.34

Chemical Identifiers

CAS 161337-67-3
Molecular Formula C4H2F12HfO13S4
Molecular Weight (g/mol) 792.758
MDL Number MFCD01321255
InChI Key XQJOFTWVKHJLHA-UHFFFAOYSA-J
Synonym hafnium iv trifluoromethanesulfonate hydrate, hafnium tetrakis trifluoromethanesulfonate , hydrate
PubChem CID 71433309
IUPAC Name hafnium(4+);trifluoromethanesulfonate;hydrate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].O.[Hf+4]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1968806
View Documents
Thermo Scientific Chemicals
L1968806
5 g
Each for $193.84
 
AAL1968814
View Documents
Thermo Scientific Chemicals
L1968814
25 g
Each for $604.34
 
Description

Description

Hafnium trifluoromethanesulfonate is a recyclable catalyst for the mononitration of o-nitrotoluene. It serves as a catalyst for homogeneous methoxycarbonylation and hydrocarboxylation reactions of phenylacetylene. It is also used as a reactant for direct Friedel-Crafts reactions of chromene hemiacetals, aminomethylation reactions under Lewis acidic conditions and prins-type cyclization reactions. It is also involved in the direct polycondensation of lactic acid. Further, it is used in cationic benzylation reactions, chemoselective thioacetalization and transthioacetalization of carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hafnium trifluoromethanesulfonate is a recyclable catalyst for the mononitration of o-nitrotoluene. It serves as a catalyst for homogeneous methoxycarbonylation and hydrocarboxylation reactions of phenylacetylene. It is also used as a reactant for direct Friedel-Crafts reactions of chromene hemiacetals, aminomethylation reactions under Lewis acidic conditions and prins-type cyclization reactions. It is also involved in the direct polycondensation of lactic acid. Further, it is used in cationic benzylation reactions, chemoselective thioacetalization and transthioacetalization of carbonyl compounds.

Solubility
Soluble in methyl cyanide, dicholomethane, dichloroethane, nitromethane, dioxane, terahydrofuran and toluene.

Notes
Incompatible with strong oxidizing agents. Moisture sensitive.
Specifications

Chemical Identifiers

161337-67-3
792.758
XQJOFTWVKHJLHA-UHFFFAOYSA-J
71433309
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].O.[Hf+4]
C4H2F12HfO13S4
MFCD01321255
hafnium iv trifluoromethanesulfonate hydrate, hafnium tetrakis trifluoromethanesulfonate , hydrate
hafnium(4+);trifluoromethanesulfonate;hydrate

Specifications

161337-67-3
C4H2F12HfO13S4
MFCD01321255
Hygroscopic
Soluble in methyl cyanide,dicholomethane,dichloroethane,nitromethane,dioxane,terahydrofuran and toluene.
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].O.[Hf+4]
792.758
774.75
Hafnium trifluoromethanesulfonate
>350°C
(CF3SO3)4Hf
5 g
hafnium iv trifluoromethanesulfonate hydrate, hafnium tetrakis trifluoromethanesulfonate , hydrate
XQJOFTWVKHJLHA-UHFFFAOYSA-J
hafnium(4+);trifluoromethanesulfonate;hydrate
71433309
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only