missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hafnium wire, 0.5mm (0.02in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 042982BY

 View more versions of this product

Catalog No. AA42982BY


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hafnium wire
7440-58-6
Hf
100 cm
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium
23986
99.95%
≈2.6g/m
0.5mm (0.02 in.) dia.
(metals basis excluding Zr), Zr nominal 3%
MFCD00011032
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
[Hf]
178.49
CHEBI:33343
Odorless
Wire

Chemical Identifiers

7440-58-6
178.49
VBJZVLUMGGDVMO-UHFFFAOYSA-N
23986
hafnium
Hf
MFCD00011032
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
CHEBI:33343
[Hf]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-166-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only