Erlab Halo Sense VOC
Halo Sense VOC monitors air quality for hazardous fumes or vapors.
Supplier: Erlab HALOVOCSENSE
Description
- Unit can be installed on the wall or placed on a lab bench
- If fugitive emissions are detected, the device alerts the user with a light and sound pulse
- Sensor type: Volatile organic compounds
- VOC Sensor replacement every 5 years
Specifications
Monitor | |
Identifying hazardous fumes or vapors |