missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktail (100X)

Prevent protein degradation and stabilize phosphorylation with this all-in-one cocktail of protease and phosphatase inhibitors.

$677.48 - $3013.81

Specifications

Product Type Protease and Phosphatase Inhibitor Cocktail
Content And Storage Upon receipt store at 4°C
Form Liquid
Product Line Halt™
Buffer Lysis Buffer
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
PI78442
missing translation for 'viewDocuments'
Thermo Scientific™
78442
24 x 100 μL
Each for $920.83
 
PI78446
missing translation for 'viewDocuments'
Thermo Scientific™
78446
10 mL
Each for $3,013.81
 
PI78440
missing translation for 'viewDocuments'
Thermo Scientific™
78440
1 mL
Each for $677.48
 
PI78444
missing translation for 'viewDocuments'
Thermo Scientific™
78444
5 x 1 mL
Each for $1,845.88
 
Description

Description

The all-in-one Thermo Scientific™ Halt Protease and Phosphatase Inhibitor Cocktail provides the convenience of a single solution with full sample protection.

Additional Information and Online Ordering

This broad-spectrum Halt Protease and Phosphatase Inhibitor Cocktail is specifically optimized to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases.The 100X cocktails, available in regular and EDTA-free formulations and several package formats, are refrigerator-stable liquids that are very easy to use for nearly any sample or experimental scale.

Highlights:

  • Complete protection all-in-one cocktail contains both protease and phosphatase inhibitors

  • Multiple package sizes choose from four convenient packages sizes to match nearly any scale

  • Convenient the 100X liquid formulation is much easier and more effective to use than individual inhibitors and other formats

  • Easy storage aqueous-based format allows for convenient cold-room storage without freezing, by contrast with DMSO-based cocktails

  • Compatible can be used in mass spectrometry (MS) because it does not contain AEBSF, which can cause peaks to shift

  • Validated fully compatible with Thermo Scientific Pierce™ Cell Lysis Buffers

Includes:

Formulations with and without supplementary vial of EDTA for metalloprotease inhibition, both of which are offered in four package sizes.

Specifications

Specifications

Protease and Phosphatase Inhibitor Cocktail
Liquid
Lysis Buffer
Protease Inhibitor, Phosphatase Inhibitor
Upon receipt store at 4°C
Halt™
Protease Inhibitor, Phosphatase Inhibitor
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.