Hamilton™ Kel-F Hub Blunt Point Needles (Luer Lock)
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. (51mm). Custom lengths and point styles are available.
$112.30 - $173.45
Specifications
Autoclavable | Not Autoclavable |
---|---|
Material | Stainless Steel |
Length (English) Needle | 2 in. |
Product Type | Kel-F Hub Needle |
Needle Point Style | Blunt |
Catalog Number | Mfr. No. | Needle Gauge | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Needle Gauge | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity | |||
14-815-576
|
Hamilton™
90512 |
12 G | 0.085 in. | 0.08 in. |
Pack of 6 Each for $112.30
|
|
|||
14-815-575
|
Hamilton™
90511 |
11 G | 0.094 in. | 0.09 in. |
Pack of 6 Each for $112.30
|
|
|||
14-815-578
|
Hamilton™
HAM90514 |
14 G | 0.062 in. | 0.062 in. |
Pack of 6 Each for $112.30
|
|
|||
14-815-596
|
Hamilton™
90536 |
10 G | 0.105 in. | 0.1 in. |
Pack of 6 Each for $112.30
|
|
|||
14-815-560
|
Hamilton™
HAM90521 |
21 G | 0.02 in. | 0.032 in. |
Pack of 6 Each for $131.09
|
|
|||
14-815-574
|
Hamilton™
HAM90134 |
22 G | 0.016 in. | 0.028 in. |
Pack of 6 Each for $131.09
|
|
|||
14-815-580
|
Hamilton™
HAM90516 |
16 G | 0.046 in. | 0.064 in. |
Pack of 6 Each for $131.09
|
|
|||
14-815-581
|
Hamilton™
HAM90517 |
17 G | 0.042 in. | 0.057 in. |
Pack of 6 Each for $131.09
|
|
|||
14-815-582
|
Hamilton™
HAM90519 |
19 G | 0.027 in. | 0.042 in. |
Pack of 6 Each for $123.09
|
|
|||
14-815-583
|
Hamilton™
HAM90520 |
20 G | 0.02 in. | 0.035 in. |
Pack of 6 Each for $131.09
|
|
|||
14-815-584
|
Hamilton™
HAM90523 |
23 G | 0.013 in. | 0.025 in. |
Pack of 6 Each for $123.09
|
|
|||
14-815-587
|
Hamilton™
HAM90525 |
25 G | 0.01 in. | 0.01 in. |
Pack of 6 Each for $131.09
|
|
|||
14-815-586
|
Hamilton™
HAM90524 |
24 G | 0.012 in. | 0.022 in. |
Pack of 6 Each for $131.09
|
|
|||
14-815-588
|
Hamilton™
HAM90528 |
28 G | 0.007 in. | 0.014 in. |
Pack of 6 Each for $173.45
|
|
|||
14-815-590
|
Hamilton™
HAM90530 |
30 G | 0.006 in. | 0.012 in. |
Pack of 6 Each for $162.85
|
|
|||
14-815-589
|
Hamilton™
HAM90529 |
29 G | 0.007 in. | 0.013 in. |
Pack of 6 Each for $162.85
|
|
|||
14-815-591
|
Hamilton™
HAM90531 |
31 G | 0.005 in. | 0.01 in. |
Pack of 6 Each for $162.85
|
|
|||
14-815-592
|
Hamilton™
HAM90532 |
27 G | 0.008 in. | 0.016 in. |
Pack of 6 Each for $162.85
|
|
|||
14-815-594
|
Hamilton™
HAM90534 |
22s G | 0.005 in. | 0.028 in. |
Pack of 6 Each for $173.45
|
|
|||
14-815-593
|
Hamilton™
HAM90533 |
26 G | 0.01 in. | 0.018 in. |
Pack of 6 Each for $123.09
|
|
|||
14-815-597
|
Hamilton™
HAM90539 |
26s G | 0.005 in. | 0.018 in. |
Pack of 6 Each for $123.09
|
|
|||
14-815-595
|
Hamilton™
HAM90535 |
18 G | 0.033 in. | 0.05 in. |
Pack of 6 Each for $133.25
|
|
|||
Description
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. (51mm). Custom lengths and point styles are available.
Kel-F Hub Compatible Syringe Terminations
- Luer Tip (LT)- The needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
- PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
- Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 10 to 31
Six needles per pack.
Specifications
Not Autoclavable | |
2 in. | |
Blunt |
Stainless Steel | |
Kel-F Hub Needle | |
Luer-tip and PTFE Luer-lock Syringes and Connectors |