Hamilton™ Small Hub Removable Needles (2.5 - 100μL Syringes)
Can be used with removable (RN)-style syringes that are 100μL and smaller.
$101.21 - $268.66
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Stainless Steel (Hub) |
Length (English) Needle | 2 in. |
Product Type | Replacement Syringe Needle |
Catalog Number | Mfr. No. | Needle Gauge | Needle Point Style | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Needle Gauge | Needle Point Style | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity | |||
14815539
|
Hamilton™
HAM778408 |
26 G | Cone | 0.01 in. | 0.018 in. |
Pack of 6 Each for $217.70
|
|
|||
14-815-541
|
Hamilton™
HAM778406 |
22 G | Cone | 0.016 in. | 0.028 in. |
Pack of 6 Each for $217.70
|
|
|||
14-815-534
|
Hamilton™
HAM775801 |
22 G | Sharp, Bevel, Curved | 0.016 in. | 0.028 in. |
Pack of 3 Each for $114.75
|
|
|||
14-815-542
|
Hamilton™
HAM778405 |
22s G | Cone | 0.0059 in. | 0.028 in. |
Pack of 6 Each for $268.66
|
|
|||
14-815-537
|
Hamilton™
HAM775803 |
22s G | Sharp, Bevel, Curved | 0.0059 in. | 0.028 in. |
Pack of 6 Each for $155.60
|
|
|||
14-815-545
|
Hamilton™
777001 |
22s G | Blunt | 0.0059 in. | 0.028 in. |
Pack of 6 Each for $145.87
|
|
|||
14-816-206
|
Hamilton™
HAM776206 |
33 G | Blunt | 0.004 in. | 0.008 in. |
Pack of 6 Each for $182.83
|
|
|||
14-816-212B
|
Hamilton™
777002 |
22 G | Blunt | 0.016 in. | 0.028 in. |
Pack of 6 Each for $106.98
|
|
|||
14-816-212A
|
Hamilton™
HAM776801 |
26s G | Blunt | 0.0051 in. | 0.018 in. |
Pack of 6 Each for $106.98
|
|
|||
14-824-9A
|
Hamilton™
775802 |
26s G | Sharp, Bevel, Curved | 0.0051 in. | 0.019 in. |
Pack of 6 Each for $114.75
|
|
|||
14-824-9B
|
Hamilton™
HAM776802 |
26 G | Blunt | 0.01 in. | 0.01 in. |
Pack of 6 Each for $101.21
|
|
|||
14-824-710
|
Hamilton™
HAM776203 |
30 G | Blunt | 0.0063 in. | 0.012 in. |
Pack of 6 Each for $138.02
|
|
|||
14-824-712
|
Hamilton™
HAM776205 |
32 G | Blunt | 0.004 in. | 0.0094 in. |
Pack of 6 Each for $138.02
|
|
|||
14-824-711
|
Hamilton™
HAM776204 |
31 G | Blunt | 0.005 in. | 0.01 in. |
Pack of 6 Each for $138.02
|
|
|||
14-824-709
|
Hamilton™
HAM776202 |
28 G | Blunt | 0.007 in. | 0.014 in. |
Pack of 6 Each for $138.02
|
|
|||
14-824-708
|
Hamilton™
HAM776201 |
27 G | Blunt | 0.0051 in. | 0.016 in. |
Pack of 6 Each for $138.02
|
|
|||
Description
- Standard needle length is 51mm (2 in.)
- Custom gauges, lengths and point styles are available
Specifications
Autoclavable | |
Stainless Steel (Hub) | |
2 in. | |
Replacement Syringe Needle |