Eisco™ Hand Centrifuge, 4 Bucket
This hand driven centrifuge can accommodate up to four 15 mL tubes. Made with die cast parts, a steel pinion, and worm drive. Designed as an economical alternative to electrical centrifuges, this centrifuge must be attached to a lab bench via the screw clamp when in use.
Supplier:  Eisco™ CH0267
Catalog No. S40585
Explore available promotions
Includes: Cetrifuge, Screw clamp, Removable crank handle, 4 plastic buckets,
Description
Description
  • Economical hand driven centrifuge
  • Accommodates four 15 mL tubes
  • Includes die cast parts, steel pinion, worm drive, 4 plastic buckets
  • Attaches to lab bench or table via screw clamp (included)
  • Removable crank handle for easy storage
Warranty
1 year limited manufacturer warranty
Specifications
Specifications
4 x 15 mL
4 buckets, hand crank, screw clamp
No
Manual
4 x 15 mL tubes