missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BRAND™ HandyStep S Wall Mount
Accessory for use with the HandyStep S
Supplier: BRAND™ 705129
Description
- For use with HandyStep S
- Accessory
- Storage rack
- Holds pipetters
Specifications
Storage Rack | |
Black | |
Polycarbonate | |
Non-sterile | |
Wall Mount |
Not Autoclavable | |
Pipetters | |
1 | |
BRAND HandyStep S |