missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hanna Instruments™ Checker™ pH Tester and Replacement Part DFS Item

For use with virtually any electrode that shares same connector

Supplier:  Hanna Instruments™ HI1270

Catalog No. 13-300-69


Explore available promotions

Description

Description

  • pH range from 0.00 to 14.00
  • Provides high accuracy with 0.01 pH resolution at 0° to 50°C
  • Accuracy is ±0.2 pH
  • 1.5V
  • RH max 95%
  • With LCD and manual two point calibration
  • 3000 hours of battery life

Specifications

Specifications

pH
Videos
SDS
Documents

Documents

Promotions

Promotions