missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BUFFER SOLUTION PH10 1/2 LT DFS Item

Used for calibration

Supplier:  Hanna Instruments CDA Inc HI7010L

Catalog No. 11-603-64


Explore available promotions

Description

Description

  • Available in different pH ranges and quantity
  • Calibrated according to NIST standards
  • With/without certificates of analysis

Specifications

Specifications

500 mL

Chemical Identifiers

0.0
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep container closed and protected from direct sunlight. Store at room temperature (+15° to +25°C).

SDS
Documents

Documents

Promotions

Promotions