Learn More
Hanna™ Instruments pH Buffer Solutions
Used for calibration
Supplier: Hanna Instruments CDA Inc HI7007M
Description
- Available in different pH ranges and quantity
- Calibrated according to NIST standards
- With/without certificates of analysis
Specifications
| 230 mL |
Chemical Identifiers
| 0.0 |
Safety and Handling
Recommended Storage : Keep container closed and protected from direct sunlight. Store at room temperature (+15° to +25°C).