missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyramex™ SL Series Hard Hats, 4-point Ratchet
Hard Hats exceed ANSI Z89.1, Type 1, Class C, G, and E for industrial head protection; CSA Z94.1-15.
$9.68 - $13.99
Specifications
Certifications/Compliance | ANSI Z89.1 |
---|---|
For Use With (Equipment) | Earmuffs, Faceshield |
For Use With (Application) | Construction |
Material | HDPE |
Product Type | Cap-style Hard Hat |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
19-161-141
|
Pyramex™
HP14160BLUE |
Blue |
Each for $11.90
|
|
|||||
19-161-137
|
Pyramex™
HP14110WHITE |
White |
Each for $13.99
Pack of 16 Each for $210.72
|
|
|||||
19-161-143
|
Pyramex™
HP14112GRAY |
Gray |
Each for $9.68
Pack of 16 Each for $145.76
|
|
|||||
Description
- Low profile design
- Tested and marked for reverse donning
- Shell constructed from High Density Polyethylene materials
- Rain trough on sides and back of helmet channel moisture away
- Soft brow pad is replaceable. Replaceable suspensions and headbands also available
- Snap Lock suspension features 'tuck away' adjustments for all day, hassle free wear
- Ratchet suspension is easy to adjust and allows the wearer to modify the fit while wearing hard hat
- Universal accessory slots fit cap mounted ear muffs and accessories. Hats also accommodate chin straps
- Testing: Impact, Penetration, Flammability, Electrical Insulation, Marking & Humidity
Specifications
ANSI Z89.1 | |
Construction | |
Cap-style Hard Hat | |
Not Vented |
Earmuffs, Faceshield | |
HDPE | |
6.5 to 8 | |
Short Brim (Cap Style) |