Hausser Scientific™ Levy™ Hemacytometer Chamber Set
This hemacytometer has an etched field with an opaque-pattern Levy chamber and double Neubauer ruling
Supplier: Hausser Scientific™ HSR3520
Description
- Complete with 2 × 0.4mm cover glasses, red blood pipet, white blood pipet, pipet filler and clear plastic case
Order Info
For replacement cover slips, see 02-671-53.
Specifications
Hemacytometer Chamber Set | |
0.1μL | |
Complete with 2 x 0.4mm cover Glasses, red Blood pipet, white Blood pipet, pipet filler and clear Plastic case |
Hausser Levy | |
Each |