missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hausser Scientific™ Howard Mold Counting Chamber
Designed for use in determining mold counts
Supplier: Hausser Scientific™ 3820
Description
Designed for use in determining mold counts
Designed for use in determining mold counts.Note: Grid is not included; sold separately.
Specifications
Hemacytometer | |
Glass | |
0.003 in. | |
Determining Mold Count |
Howard Mold. (Grid is sold separately.) | |
Each | |
0.01cm | |
Cover slips |