missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Haws™ Wall Mount Eye/Face Wash and Body Spray Head with Recoil Hose

Haws™ Wall Mount Eye/Face Wash and Body Spray Head with Recoil Hose features an inverted flow design consistent with all EMT, emergency room and doctors' office protocols

Supplier:  Haws™ 8905

Catalog No. 19-033-735


Explore available promotions

Description

Description

  • Eyewash streams provide Zero Vertical Velocity™ stream engineering with even laminar flow for enhanced comfort, stability and effectiveness.
  • Dust cover allows for the 8905 to stay protected from debris when not in use
  • Easily detected and maintained
  • Equipped with safety sign and test card to record weekly testing
  • 12-foot (365.8cm) yellow recoil hose provides an extended coverage area to further optimize the irrigation process
  • In order to comply with state codes the use of a vacuum breaker is required with all hand held eye/face wash body spray units (Haws offers model SP212)

Warranty

1 year from ship date (Limited)

Specifications

Specifications

Squeeze Handle
Green, Yellow
14.75 in.
Body Spray Head; 12-foot (365.8cm) Yellow Recoil Hose
Rubber
0.5 in.
ANSI Z358.1
3.7 gpm
37.465 cm
1.27 cm
Wall
200 psi
Videos
SDS
Documents

Documents

Promotions

Promotions