missing translation for 'onlineSavingsMsg'
Learn More

Hayem Diluting Fluid, for Erythrocyte (Red Cell) Count, Ricca Chemical

For Erythrocyte (Red Cell) Count

$104.75 - $104.75

Specifications

Chemical Name or Material Hayem Diluting Fluid
Physical Form Liquid
CAS 7732-18-5,7757-82-6,7647-14-5,7487-94-7
CAS Min % 95.0,2.37,0.47,0.24
CAS Max % 99.0,2.47,0.49,0.25
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
3500-16
View Documents
Ricca Chemical Company
350016
500 mL Amber Glass Bottle
Each for $104.75
Only null left
 
Description

Description

Specifications

Specifications

Hayem Diluting Fluid
7732-18-5,7757-82-6,7647-14-5,7487-94-7
99.0,2.47,0.49,0.25
Colorless
Liquid
95.0,2.37,0.47,0.24
Amber Glass Bottle
Laboratory
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'