missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Hazardous Material Label: BLEACH
Provide workers with all of the chemical information needed for hazardous materials they may encounter. Hazardous Material Labels are made of vinyl and display text in black on yellow.
Supplier: Brady™ 93496
Description
- Provides workers with chemical information of hazardous materials
- Made from B-928 vinyl material
- Label Size: W x H: 4.5 x 3.75 in.
Specifications
Not Autoclavable | |
Label | |
Black, Yellow | |
Vinyl (B-928) | |
Hazardous Materials Label | |
BLEACH | |
Pack of 25 Labels | |
25 Pk. | |
4.5 x 3.75 in. (W x H) | |
3.75 x 4.5 in. | |
114.30 mm | |
95.25 mm |
Globally Harmonized System of Classification and Labeling of Chemicals (GHS), NFPA 704, OSHA 1910.1200 | |
Cold-temperature, Permanent Acrylic, Cold-temperature | |
Label | |
Single-side Printable | |
Non-sterile | |
Facility Identification, Hazard Communication Identification | |
English | |
Rectangle | |
0.0049 in. | |
4.5 in. | |
3.75 in. |