missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Hazardous Material Label: DIETHYL ETHER
Provide workers with all of the chemical information needed for hazardous materials they may encounter. Brady™ Hazardous Material Labels are made of vinyl and display text in black on yellow.
Supplier: Brady™ 93524
Description
- Provides workers with chemical information of hazardous materials
- Made from B-928 vinyl material
- Label Size: W x H: 4.5 x 3.75 in.
Specifications
Acrylic, Permanent, Cold-temperature | |
Vinyl (B-928) | |
DIETHYL ETHER | |
Rectangle | |
0.0049 in. |
Black, Yellow | |
Hazardous Materials Label | |
25/Pk. | |
4.5 x 3.75 in. (W x H) |