missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Hazardous Waste Label: NON-HAZARDOUS WASTE
Identify hazardous waste materials for transportation off-site. Brady™ Hazardous Waste Labels are made of vinyl and comply with all current federal regulations regarding identification of hazardous waste material transportation.
$61.47 - $109.79
Specifications
Adhesive Type | Acrylic, Permanent |
---|---|
Color | White, Green |
Material | Vinyl (B-7569) |
Product Type | Hazardous Waste Label |
Display Text | NON-REGULATED WASTE |
Catalog Number | Mfr. No. | Size (English) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size (English) | Price | Quantity | |||||
19-110-974
|
Brady™
121154 |
6 x 6 in. |
Pack of 100 Each for $109.79
|
|
|||||
19-110-978
|
Brady™
121159 |
6 x 6 in. |
Pack of 50 Each for $61.47
|
|
|||||
Description
- To identify containers, cylinders and drums holding hazardous waste
- Made of B-7569 vinyl
- Available in size (W x H): 15.2 x 15.2cm (6 x 6 in.)
Specifications
Acrylic, Permanent | |
Vinyl (B-7569) | |
NON-REGULATED WASTE | |
6 x 6 in. |
White, Green | |
Hazardous Waste Label | |
Square | |
0.004 in. |