Thermo Scientific™ HCS Studio™ 2.0 Cell Analysis Software, client installation
Acquire and analyze cellular imaging and data with a premier high content instrument control, visualization, and analysis software.
Supplier: Thermo Scientific™ SX000041A
Description
Provide control, visualization, and analysis, using the Thermo Scientific™ HCS Studio™ 2.0 Cell Analysis Software, a premier high content instrument software integrating quantitative cell analysis, autofocus, autoexposure, and cell subpopulations analysis tools with simple plate scanning.
A Measurable Difference for Everyone:
Workflow-oriented and simple to navigate, HCS Studio 2.0 Cell Analysis Software allows access to all instrument configuration and control functions with intuitive tools. Guided workflows with visual feedback allow for assay protocol setup, image acquisition, analysis, and scanning. Save time by acquiring and analyzing simultaneously. By ensuring only the statistically significant number of cells is captured, less time is spent scanning unnecessarily.
HCS Studio 2.0 Cell Analysis Software expertly balances flexibility with ease-of-use through intuitive interfaces, intelligent design and powerful image analysis tools leading to meaningful dynamic measurements at the cell-, well- and field-level.Highlights:
- Analyze Multiple Image Formats – Allows image formats from a variety of sources and in several different file types to be imported singly or in batch, organized for analysis, and then analyzed and stored in Thermo Scientific™ Store™ Image and Database Management Software
- Compare and Optimize Two Biological Extents – Allows you to apply and interact with two biology types simultaneously without the risk of bias
- Scale for Complex Assays – Scales seamlessly providing a host of algorithmic, visualization, and online help tools reducing the complexity and time investment of developing assays
Specifications
HCS Studio | |
ArrayScan High Content Analysis (HCA) Readers, CellInsight CX5 High Content Screening (HCS) Platform | |
Image Acquisition, Image Analysis | |
1 each |
For Research Use Only. Not for use in diagnostic procedures.