Fisherbrand™ HDPE, Rectangular Carboy with Spigot
For Use With: Ideal for storing and dispensing lab water and reagents; Color: White
 View more versions of this product
Catalog No. 03-007-643
Explore available promotions
Description
Description
Carboys Ideal for storing and dispensing lab water and reagents.
Ideal for storing and dispensing lab water and reagents
  • Durable construction for long service life
  • Molded graduations for 1 gallon and 51 increments
  • Choice of materials for general use or autoclavability
  • Space-saving rectangular design
  • Wide mouth closure allows for easy filling and cleaning access
  • Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
  • Spigot spout without the adapter fits 5/8-in. ID tubing
  • Materials meet FDA requirements for food use and USP VI criteria (non-toxic)
Specifications
Specifications
Not Autoclavable
9 L
White
HDPE
Carboy with Spigot
2.4 gal.
FDA, USP VI
Ideal for storing and dispensing lab water and reagents
Wide
Rectangular