Supertek™ Heat Conductometer, Five Spoke
A Heat Conductometer is used for comparing the thermal conductivity of five metals: aluminum, brass, steel, nickel, and copper. The 5 spokes radiate from a brass hub. Each rod is marked on the rods.
Supplier: Supertek™ PH20278
Description
- A Heat Conductometer is used for comparing the thermal conductivity of five metals.
- Five metals: aluminum, brass, steel, nickel, and copper.
- The 5 spokes radiate from a brass hub.
- Each rod is marked on the rods.
- There are notches in each rod to hold the wax.
- Comes assembled with a supply of wax.
- Instructions included
Specifications
Heat Conductometer | |
6+ | |
Heat Conductomer, Wax | |
1 Lab Group |