Fisherbrand™ Heat/Cool Thermal Mixer II PROMO
For precise sample preparation when working with DNA, RNA, plasmids, cultures and enzymatic reactions.
$9298.53 - $9465.98
Specifications
Electrical Requirements 12 VDC via 115/230 VAC adapter
Product Type Thermal Mixer
Depth (English) 11 in.
Depth (Metric) 27.94 cm
Height (English) 7.5 in.
View More Specs
Products 9
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
11-700-167
View Documents
Fisherbrand™
270800FPROMO12
Conical Tube Blocks (15-600-333 and -334)
Each for $9,465.98
11-700-168
View Documents
Fisherbrand™
270800FPROMO13
Conical Tube Blocks (15-600-333 and 15-600-335)
Each for $9,298.53
11-700-169
View Documents
Fisherbrand™
270800FPROMO14
Conical Tube Blocks (15-600-333 and 15-600-336)
Each for $9,298.53
11-700-170
View Documents
Fisherbrand™
270800FPROMO15
Conical Tube Blocks (15-600-333 and 15-600-337)
Each for $9,298.53
11-700-171
View Documents
Fisherbrand™
270800FPROMO16
Conical Tube Blocks (15-600-333 and -338)
Each for $9,298.53
11-700-172
View Documents
Fisherbrand™
270800FPROMO17
Conical Tube Blocks (15-600-333 and 15-600-339)
Each for $9,298.53
11-700-173
View Documents
Fisherbrand™
270800FPROMO18
Conical Tube Block (15-600-333) and Standard/Deep Microplate Block (15-600-340)
Each for $9,298.53
11-700-174
View Documents
Fisherbrand™
270800FPROMO19
PCR Plate Blocks (15-600-341)
Each for $9,298.53
11-700-175
View Documents
Fisherbrand™
270800FPROMO20
Conical Tube Block (15-600-333) and PCR Plate Block (15-600-341)
Each for $9,298.53
Description
Description
  • Large color touch screen
  • Interchangeable blocks
  • Auto-recognition of blocks
  • Mixing and temperature control for small samples
  • For laboratory use
  • Designed by scientists for scientists
  • Easy to use and program
Intuitive menu navigation
  • Home display indicates step number and program name
  • Unit operating status is indicated by bright red text
  • “Stop, Play, Pause, Skip and Quick Mix” icons are easy to use
  • Instantly change parameters for one step
  • Turn off or on parameters as needed during process
  • Minimum and maximum parameters are automatically determined by block type
  • View and modify all steps on program screen
  • Automatically repeat one sequence as many times as needed
  • Use as many as steps as needed, up to five available
  • Easily select, launch, change, and save programs
  • Programs can have long text names for easy process recall
  • Turn on parameter lock to protect important processes
  • Stores up to 25 programs
Note: Other block options are available for regular tubes, cryo tubes, PCR plates and more.
Specifications
Specifications
12 VDC via 115/230 VAC adapter
11 in.
7.5 in.
7 in.
Thermal Mixer
27.94 cm
19.05 cm
17.78 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'