Learn More
Keystone Cleanroom Products Heat-Seal Tyvek Autoclave Bag
Keystone Cleanroom Products Heat-Seal Tyvek Autoclave Bag is a portable isolator in an easy to use, economical bag form. Ideal for difficult-to-sterilize equipment that typically requires aseptic connections after sterilization.
Supplier:  Keystone Cleanroom Products BGT2A1626MITT2WOCPE
 View more versions of this product
Catalog No. 19-990-124
Only null left
Explore available promotions
Description
Description
  • Tyvek™/PE Film construction for excellent strength and exceptional microbial barrier
  • Designed for steam sterilization
  • Heat Seal Closure
  • Integrated Mittens
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Make aseptic connections inside the Isolator Bag
  • Eliminates risks of contamination
Specifications
Specifications
Autoclavable
COC
White
Steam Sterilization
Tyvek
Yes
26 x 16 in.
66 cm
41 cm
35,963 mL
Heat Seal
Single-use
Integrated mittens, See-through window, and Steam indicator
Non-sterile
Steam
26 in.
16 in.
Autoclave Bag
Product Suggestions
Product Suggestions