Eisco™ Heavy Duty Boss Head, Black Coated, up to 20mm rods
This heavy duty boss head is die casted alloy with a black coat finish. It accepts rods up to 20mm in diameter at a right angle to one another. Large thumb screw for adjustments.
Supplier:  Eisco™ CH0665B
Catalog No. S24268
Explore available promotions
Description
Description
Specifications
Specifications
Adjustable Clamp
5 in.
5 cm
Fixed
Zinc Die Cast, Plastic Coated
2 in.
Black
Product Suggestions
Product Suggestions