missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heidolph™ 3870 ELVC Tuttnauer™ Vertical Autoclave

Top loading autoclaves with advanced microprocessor control panel and chamber made of 316Ti stainless steel

Supplier:  Heidolph™ 023210950

Catalog No. 13-889-564

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Includes: Internal Thermometer, Water-sensing Electrode, External Pressure Gauge, Sample Temberature Probes, Printer, USB, Wire Baskets (set of 2)

Description

Description

Advanced Control system

  • F0 software control protects liquid media by minimisng exposire to high temperatures during sterilization
  • PID (Proportional Integral Differential) pressure control
  • Stores last 200 cycles in built-in memory
  • Up to 6 temperature sensors and 4 pressure sensors can be connected
  • 30 Identification codes and passwords for access level control
  • Controller and software comply with the 21 CFR part 11 standard
  • In/Out test (enables technical to check each system component separately)
  • Sterilization Temperature range 105° to 137°C
  • Optional Disinfection/Isothermal Temperature Range 60° to 105°C
  • Optional Independent Recording for cross-checking cycle measurements
  • Filter replacement notifications based on number of cycles
  • Fast Liquid Cooling
Multi-Color Display
  • Enables easy reading
  • Color indicates cycle stage
  • Built-in view of historical cycle data
  • Graphical display of Temperature and Pressure
  • Gives quick access to important information
  • Easily operated in mulitple languages

Applications:

For laboratories in research institutes, universities, pharmaceutical, food, chemical and biotechnological industries

Specifications

Specifications

20 A
85 L
230V/ 50-60 Hz, 3-phase
230 V
“Plug & Play” to operate only with an electric power connection. No need to connect autoclave to building utilities for distilled water, tap water and drain.
Autoclave, Vertical
50/60 Hz 3 Phase
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions