missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heidolph™ Reactor-Ready Duo Lab Reactor
Supports two jacketed glass reaction vessels of different or similar volume from 100mL to 5L
Supplier: Heidolph™ 015970100
Description
- Reactor allows rapid exchange of both vessels independently, with quick-release vessel clamp and hose couplings
- Choice of manifold kits allow two vessels to run from single thermo regulator or two separate thermo regulators
- System accepts two overhead stirrers which can be operated and moved independently
- Robust heavy-duty framework features solid stainless steel support rods
- Two separate quick-release damps support each vessel independently
- Large drip tray retains vessel contents
Manifold and Hose kits much be ordered separately
- Parallel synthesis or reaction optimization: Use similar or different size vessels and vary stirring speed, stirrer shape and temperature between vessels
- Two stage reaction: Transfer reactant from one vessel to other using vacuum or pump
- Single reaction vessel: Using second vessel as either receiving or feed vessel (where reagents can be pre-heated or pre-cooled prior to addition)
- Use optional Control Software to control fluid transfer between vessels
Specifications
Parallel synthesis or reaction optimization | |
Reactor-Ready Duo Lab Reactor |