Heidolph™ Overhead Stirrer Impellers - VISCO JET
Visco Jet Impellers are recommended for larger quantities of high viscosity media and gel. Fields of use include food and beverage production, cosmetics, paint production, etc.
Supplier: Heidolph™ 036300500
Description
The VISCO JET Mixing System is the result of the so-called cone principle. Turbulent flows are created even at low speeds at the taper end by acceleration, displacement and retardation. These flows advance through the stirred medium and result in the new dynamic mixing motion.
- Reduce your process times significantly while performing the best mixing results
- One system for literally all mixing tasks for low to high-viscosity media
- Even with high-viscosity media and gels which naturally do not mix when common impellers are used you will observe an immediate flow through the entire beaker
- This technology allows for de-gassing of gels while preventing air intake and foaming
Fields of use: Beverage production, dairy products, food, sugar & candy production, chemistry/petro chemistry, ceramics, water treatment, cosmetics, colorant/paint production and paper manufacture, etc.
Specifications
4.7 in. | |
12 cm | |
17 to 30 cm dia. Vessel | |
19.6 in. | |
120 to 500 rpm | |
64 oz. | |
4 lb. |
0.39 in. | |
1 cm | |
Mixing larger quantities of high viscosity liquid and gel | |
500 mm | |
VISCO Jet - 120 Steel | |
Stainless Steel |