Heidolph™ Hei-TORQUE Overhead Stirrer Stands
Hei-TORQUE Overhead Stirrer Stand for use with Hei-TORQUE Overhead Stirrers.
Supplier:  Heidolph™ 036300540
 View more versions of this product
Catalog No. 13-880-148
Explore available promotions
Description
Description
  • Available in a variety of lengths
Specifications
Specifications
1.25 in.
Hei-TORQUE Core, Hei-TORQUE Expert, Hei-TORQUE Ultimate
725 to 1025 mm
Overhead Stirrer Stand
22 lb.
3.2 cm
28.5 to 40.35 in.
Telescope
10 kg