Heidolph™ Tornado Plus Overhead Stirring System, Stirrer Shafts
Stirrer shafts for use with Heidolph Tornado Plus
Supplier:  Heidolph™ 015861840
 View more versions of this product
Catalog No. 05-403-865
Explore available promotions
Description
Description
A range of PTFE stirrer paddles have been designed specifically for the Tornado.
  • Centrifugal
  • Anchor
  • Propeller
The Centrifugal stirrer paddles will fit into any one of the standard narrow neck Carousel 6 Plus 50 mL, 100 mL, 170 mL or 250 mL flasks. The Anchor and Propeller stirrer paddles have been designed to fit in the wide neck 100 mL and 250 mL flasks.
Specifications
Specifications
Fits 100 and 250mL wide-neck flasks
6
Propeller Stirrer Shaft