Heidolph™ CHAMBER BRITE Sterilizer Cleaner
Keeps sterilizer clean and free of water deposit buildup. Heidolph CHAMBER BRITE Sterilizer Cleaner is specially designed for steam sterilizers.
Supplier: Heidolph™ 023212803CA
Description
- Fast, convenient and effective way to clean and descale steam sterilizers, including chamber, reservoir and tubing
- Convenient, premeasured packets replace messy liquids, measuring and scrubbing
- Simply sprinkle packet into chamber and run
- Regular use optimizes autoclave function and extends operating life
Specifications
Clean and descale steam sterilizers | |
Tuttnauer benchtop sterilizer | |
Sterilizer Cleaner |