3M™ Versaflo™ M-Series Helmet Welding Knob and Pivot Kit
Allows the faceshield to pivot when raised, lowered or repositioned. 3M™ Versaflo™ M-Series Helmet Welding Knob and Pivot Kit is a complete replacement of the welding knob, internal, and external pivot joints.
Supplier: 3M™ 7100168648
Description
- Weld shield can be installed with common hand tools
- Available as replacement parts
- User is able to lift weld shield independently from respiratory visor
- Used for attaching Speedglas™ M-153SG attachment to Versaflo™ M-400 Series helmets
- 3M Catalog# M-146SG
Specifications
ANSI Z89.1 Type I, Class G | |
3M Versaflo Respiratory M-Series Helmet Assembly | |
M-146SG | |
1 Ea. |
Black | |
Welding | |
Accessories and Spare Parts | |
Standard |