missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hematein, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 370840250

 View more versions of this product

Catalog No. AC370840250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hematein
180°C
Authentic
C16H12O6
25 g
15,4669
Solubility in water: very slightly soluble. Other solubilities: slightly solubel in ethanol,ether,soluble in alkaline solutions,insoluble in benzene,chloroform
OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O
300.27
300.26
475-25-2
Brown
Glass bottle
MFCD00036187
18,IV,3343
hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein
HLUCICHZHWJHLL-UHFFFAOYNA-N
3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one
10138
Crystalline Powder

Chemical Identifiers

475-25-2
300.27
HLUCICHZHWJHLL-UHFFFAOYNA-N
10138
OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O
C16H12O6
MFCD00036187
hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein
3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-492-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only