Thermo Scientific™ Hematocrit Fixed-Angle Rotor
Save time in clinical processing applications with a fixed-angle rotor that provides dedicated blood processing to achieve 24 capillary separations per run.
Supplier: Thermo Scientific™ 75005733
Description
Save time in clinical processing applications with the Thermo Scientific™ Hematocrit Fixed-Angle Rotor, which provides dedicated blood processing to achieve 24 capillary separations per run.
Achieve red blood cell capillary separations.
Result reading chart provided.
Secure, push-button Auto-Lock rotor exchange in as little as 3 seconds delivers:
- Trouble-free rotor installation and removal
1. No tools are required
2. No complicated rotor changing procedures
- Clear chamber access for cleaning convenience
- Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
Ordering Information:
Includes linear scale. Replacement capillaries (75000923), sealing kit (75000964) and replacement linear scale (75000938) are sold separately. Contact your sales representative for more information.
Specifications
Hematocrit Fixed-Angle Rotor | |
24 Capillary Tubes | |
13,300 rpm | |
1559 | |
90° |
Fixed Angle Rotor | |
17,860 x g | |
85 mm | |
Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges |